Interested in a bulk or custom version of this product? Request pricing.

IL1RAPL2 (IL1R9) Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10156H02H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin-1 receptor accessory protein-like 2 (IL1RAPL2/IL1R9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in cell biology research applications. It can also be used in molecular biology applications as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 356) of human IL1RAPL2 precursor (NP_059112.1) fused to the Fc region of human IgG1 at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in 100 MM Glycine, 10 MM NaCl , 50 MM Tris , pH 7.5.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Gene Regulation, Genetic Diseases, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IL1R9, IL-1R9, TIGIRR-1, IL1RAPL-2
Gene ID: 26280
Gene Symbol: L1RAPL2
Molecular Weight: 66 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Cytokines & Receptors
Protein Form: Recombinant
Protein Name: Interleukin 1 receptor accessory protein-like 2
Protein Subtype: Cytokine Receptors, Interleukins
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products