Interested in a bulk or custom version of this product? Request pricing.

IL1RAPL2 Recombinant Human Protein, His Tag

Catalog number: 10156H08H50

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 1 receptor accessory protein-like 2 (IL1RAPL2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 356) of human IL1RAPL2 (NP_059112.1) fused to a polyhistidine tag at the C-terminus..

Activity: Measured by its ability to inhibit IL1B dependent proliferation in D10.G4.1 mouse helper T cells.
Approximately 5-20 µg/ml will inhibit 50% of the biological response due to 40 pg/ml of IL1B.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Gene Regulation, Immunology, Neurobiology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IL1R9, IL-1R9, TIGIRR-1, IL1RAPL-2
Gene Symbol: IL1RAPL2
Molecular Weight: 40.7 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Cytokines & Receptors
Protein Name: interleukin 1 receptor accessory protein-like 4
Protein Subtype: Cytokine Receptors, NFκB Pathway Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 95% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products