Interested in a bulk or custom version of this product? Request pricing.

S100B Recombinant Human Protein, N-His Tag

Catalog number: 10181H07E250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

S100 Calcium Binding Protein B (S100B), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) fused to a polyhistidine tag at the N-terminus.

Activity:
1. Measured by its ability to bind human AGER in a functional ELISA.
2. Measured by its ability to bind TP53 in a functional ELISA.

Formulation: PBS, 0.2 µg/µl BSA, 10% glycerol, pH 7.5.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: NEF, S100, S100beta
Gene Symbol: S100B
Molecular Weight: 12 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Name: S100 calcium binding protein B
Protein Subtype: Neurotrophic Factors, Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Neurobiology, Other Research, Signal Transduction
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols