Sterilin™ Special Petri Dishes
Actual product may vary
Thermo Scientific™
Sterilin™ Special Petri Dishes
Perform nonstandard applications with Thermo Scientific™ Sterilin™ Special Petri Dishes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
101VREDRed
101VBLUEBlue
502VFClear
Catalog number 101VRED
Price (EUR)
-
Color:
Red
Perform nonstandard applications with Thermo Scientific™ Sterilin™ Special Petri Dishes. Minimum order quantities apply.
Ordering Information:
Product not available in all geographic regions.
Specifications
Certifications/Compliance90mm dishes tested in accordance with EN ISO 24998:2008 standard
No. per Case500
No. per Pack20
ShapeRound
TypePetri Dish
Vent3
Bottom ShapeFlat
ColorRed
Diameter (Metric)90 mm
Height (Metric)16.2 mm
SterilityAseptically Manufactured
Surface TreatmentNon-treated
Unit SizeEach