Interested in a bulk or custom version of this product? Request pricing.

ERBB3 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag

Catalog number: 10201H02H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

V-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (ERBB3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies..

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 643) of human ERBB3 precursor (NP_001973.2) fused to the Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized human ERBB3-Fc at 1 µg/well can bind biotinylated recombinant human NRG1 with a linear range of 0.032-0.16 µg/ml.

Formulation: Lyophilized in PBS, pH7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Growth Factors & Receptors
Research Category: Cardiovascular Research, Oncology, Signal Transduction, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: HER3, LCCS2, ErbB-3, c-erbB3, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3
Gene ID: 2065
Gene Symbol: ERBB3
Kinase Family: EGF Receptor (ErbB) Kinase Family
Molecular Weight: 95.4 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: v-erb-b2 erythroblastic leukemia viral oncogene homolog 3
Protein Subtype: Growth Factor Receptors, Receptor Tyrosine Kinases
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products