Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
10208-1-AP is a rabbit polyclonal antibody raised against the N-terminal of human EFTUD2.
Immunogen sequence: IGPELDSDE DDDELGRETK DLDEMDDDDD DDDVGDHDDD HPGMEVVLHE DKKYYPTAEE VYGPEVETIV QEEDTQPLTE PIIKPVKTKK FTLMEQTLPV TVYEMDFLAD LMDNSELIRN VTLCGHLHHG KTCFVDCLIE QTHPEIRKRY DQDLCYTDIL FTEQERGVGI KSTPVTVVLP DTKGKSYLFN IMDTPGHVNF SDEVTA (13-217 aa encoded by BC002360)
EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 116 kDa U5 small nuclear ribonucleoprotein component; Elongation factor Tu GTP-binding domain-containing protein 2; hSNU114; KIAA0031; SNU114 homolog; U5 small nuclear ribonucleoprotein 116 kDa; U5 snRNP-specific protein, 116 kDa; U5-116 kDa; U5-116KD; unnamed protein product
Gene Aliases: 116kDa; EFTUD2; KIAA0031; MFDGA; MFDM; SNRNP116; SNRP116; Snu114; U5-116KD
UniProt ID: (Human) Q15029, (Mouse) O08810
Entrez Gene ID: (Human) 9343, (Rat) 287739, (Mouse) 20624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support