Interested in a bulk or custom version of this product? Request pricing.

BMP5 Recombinant Human Protein, N-hIgG1-Fctag

Catalog number: 10214H01H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Bone Morphogenetic Proteins 5 (BMP5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the carboxy-terminal domain (Gln 324-His 454) (the mature chain) of human BMP5 (NP_066551.1) fused to Fc region of human IgG1 at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in 50 mM Tris, 100 mM Glycine, 10 mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cardiovascular Research, Immunology, Neurobiology, Oncology, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: BMP5, MGC34244
Gene ID: 653
Gene Symbol: BMP5
Molecular Weight: 41.5 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Growth Factors & Receptors
Protein Form: Full Length, Recombinant
Protein Name: Bone morphogenetic proteins 5
Protein Tag or Fusion: IgG1 Fc Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols

Recommended products