Acenaphthenequinone, 95%
Acenaphthenequinone, 95%
Acenaphthenequinone, 95%
Thermo Scientific Chemicals

Acenaphthenequinone, 95%

CAS: 82-86-0 | C12H6O2 | 182.18 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 102190250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS82-86-0
IUPAC Name1,2-dihydroacenaphthylene-1,2-dione
Molecular FormulaC12H6O2
InChI KeyAFPRJLBZLPBTPZ-UHFFFAOYSA-N
SMILESO=C1C(=O)C2=C3C1=CC=CC3=CC=C2
さらに表示
Appearance (Color)Yellow to ochre
Infrared spectrumConforms
HPLC>=94.0 %
Appearance (Form)Powder and/or granules
Melting point253°C to 263°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only