Acethydrazide, 95%
Acethydrazide, 95%
Acethydrazide, 95%
Thermo Scientific Chemicals

Acethydrazide, 95%

CAS: 1068-57-1 | C2H6N2O | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 102350250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1068-57-1
IUPAC Nameacetohydrazide
Molecular FormulaC2H6N2O
InChI KeyOFLXLNCGODUUOT-UHFFFAOYSA-N
SMILESCC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on drying(50°C, 3 h) Balance
Appearance (Form)Moist crystals and/or chunks
Assay>=94 % (on dry substance)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only