Interested in a bulk or custom version of this product? Request pricing.

CD40LG (CD154) Recombinant Human Protein

Catalog number: 10239H08E250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD40 Ligand (CD40LG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the soluble form of human CD40LG (NP_000065.1) (Glu 108-Leu 261) fused to an N-terminal Met.

Activity: Measured by its binding ability in a functional ELISA:
1. Immobilized human CD40 at 2 µg/ml (100 µl/well) can bind biotinylated human CD40LG with a linear range of 15.6-500 ng/ml.
2. Immobilized human CD40 / Fc at 2 µg/ml (100 µl/well) can bind biotinylated human CD40LG with a linear range of 7.8-125 ng/ml.

Formulation: PBS, 0.2 µg/µl BSA, 10% glycerol, pH7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Signal Transduction
Expression: E. coli
Form: Frozen
Gene Aliases: RP3-527F8.3, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L
Gene Symbol: CD40LG
Molecular Weight: 17.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Name: CD40 ligand
Protein Subtype: Other Proteins, TNF (Tumor Necrosis Factor)
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols