2-Acetylbutyrolactone, 99+%
2-Acetylbutyrolactone, 99+%
2-Acetylbutyrolactone, 99+%
Thermo Scientific Chemicals

2-Acetylbutyrolactone, 99+%

CAS: 517-23-7 | C6H8O3 | 128.13 g/mol
数量:
100 mL
250 mL
1 L
製品番号(カタログ番号) 102492500
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS517-23-7
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4580 to 1.4610 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only