Acetylenedicarboxylic acid, 98%
Acetylenedicarboxylic acid, 98%
Acetylenedicarboxylic acid, 98%
Thermo Scientific Chemicals

Acetylenedicarboxylic acid, 98%

CAS: 142-45-0 | C4H2O4 | 114.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 102520250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS142-45-0
IUPAC Namebut-2-ynedioic acid
Molecular FormulaC4H2O4
InChI KeyYTIVTFGABIZHHX-UHFFFAOYSA-N
SMILESOC(=O)C#CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=97.5 %
Appearance (Color)White to cream to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only