Interested in a bulk or custom version of this product? Request pricing.

FCGR2B (CD32b) Recombinant Human Protein, His Tag

Catalog number: 10259H08H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fc gamma receptor II-b (FCGR2B/CD32b), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Ala 46-Pro 217) of mature human FCGR2B (NP_001002274.1) protein was expressed with a N-terminal signal peptide and a C-terminal polyhistidine tag.

Activity: Measured by its binding ability in a functional ELISA . Immobilized recombinant human FCGR2B at 10 µg/ml(100 µl/well) can bind human IgG2 with a linear range of 0.16-6.4 µg/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins, Growth Factors & Receptors
Research Category: Immunology, Inflammation
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CD32, CD32B, FCG2, FCGR2, FCGR2B, IGFR2
Gene ID: 2213
Gene Symbol: FCGR2B
Molecular Weight: 20.8 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: Fc fragment of IgG, low affinity IIb, receptor (CD32)
Protein Subtype: Fc Receptors, Glycoproteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols