2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
Thermo Scientific Chemicals

2-Acetylpyridine, 98%

CAS: 1122-62-9 | C7H7NO | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 102685000
Price (USD)
496.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
496.00
Each
Chemical Identifiers
CAS1122-62-9
IUPAC Name1-(pyridin-2-yl)ethan-1-one
Molecular FormulaC7H7NO
InChI KeyAJKVQEKCUACUMD-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light brown
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5200 to 1.5250 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only