Acrolein diethyl acetal, 94%
Acrolein diethyl acetal, 94%
Acrolein diethyl acetal, 94%
Thermo Scientific Chemicals

Acrolein diethyl acetal, 94%

CAS: 3054-95-3 | C7H14O2 | 130.19 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
產品號碼 102771000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS3054-95-3
IUPAC Name3,3-diethoxyprop-1-ene
Molecular FormulaC7H14O2
InChI KeyMCIPQLOKVXSHTD-UHFFFAOYSA-N
SMILESCCOC(OCC)C=C
檢視更多
規格Specification Sheet規格表
Refractive index1.3970 to 1.4030 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=92 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only