Adenosine 5'-monophosphate, 99%
Adenosine 5'-monophosphate, 99%
Adenosine 5'-monophosphate, 99%
Thermo Scientific Chemicals

Adenosine 5'-monophosphate, 99%

CAS: 61-19-8 | C10H14N5O7P | 347.22 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 102790250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS61-19-8
IUPAC Name{[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Molecular FormulaC10H14N5O7P
InChI KeyUDMBCSSLTHHNCD-YPLCUDRINA-N
SMILESNC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
檢視更多
規格Specification Sheet規格表
Appearance (Form)Crystalline powder
UV transmittance>=95.0 % (c=1, 0.2 N NaOH, 430 nm) 1 cm cell
Infrared spectrumConforms
Loss on drying=<6 % (105°C, 5 hrs)
Ratio of Absorbances0.76 to 0.80 (A250/A260)
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only