Adenosine 5'-triphosphate, disodium salt hydrate, 98%
Adenosine 5'-triphosphate, disodium salt hydrate, 98%
Adenosine 5'-triphosphate, disodium salt hydrate, 98%
Thermo Scientific Chemicals

Adenosine 5'-triphosphate, disodium salt hydrate, 98%

CAS: 34369-07-8 | C10H14N5Na2O13P3 | 551.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
10280050050 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 102800500
Price (USD)
666.65
Special offer
Online exclusive
Ends: 28-Dec-2026
784.00
Save 117.35
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
666.65
Special offer
Online exclusive
Ends: 28-Dec-2026
784.00
Save 117.35
Each
Chemical Identifiers
CAS34369-07-8
IUPAC Namedisodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC10H14N5Na2O13P3
InChI KeyTTWYZDPBDWHJOR-WCYUCLFNNA-L
SMILES[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
UV spectrophotometry98 to 102 % (lambda max. 259 nm, pH 7)
Water=<10.0 % (K.F.)
pH2.5 to 3.5 (5 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only