Allyl Alcohol, 99%, extra pure
Allyl Alcohol, 99%, extra pure
Allyl Alcohol, 99%, extra pure
Thermo Scientific Chemicals

Allyl Alcohol, 99%, extra pure

CAS: 107-18-6 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 102860025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-18-6
IUPAC Nameprop-2-en-1-ol
Molecular FormulaC3H6O
InChI KeyXXROGKLTLUQVRX-UHFFFAOYSA-N
SMILESOCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Infrared spectrumConforms
GC>=98.5 %
Acidity (CH3COOH)=<0.02 %
Water=<0.3 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only