Sterilin™ 100mm Square Petri Dishes
Thermo Scientific™

Sterilin™ 100mm Square Petri Dishes

Conduct various applications with Thermo ScientificTM SterilinTM 100mm Square Petri Dishes, available in both compartmentalized and non-compartmentalized formats.
Have Questions?
Catalog NumberWell CountVent
10325Selectable
Catalog number 103
Price (USD)
-
Well Count:
25
Vent:
Selectable

Conduct various applications with Thermo Scientific™ Sterilin™ 100mm Square Petri Dishes, available in both compartmentalized and non-compartmentalized formats. Manufactured in Clean Room Class 7 Conditions to exclude microbiological contamination.

NOT AVAILABLE IN AMERICA

Non-compartmentalized:

  • Ideal for antibiotic sensitivity testing when a large surface area and very flat base is required

Compartmentalized:

  • Ideal for small volume liquid media work or for sample storage
  • 25 compartments each have a surface area of 3.7cm2 and a capacity of 5mL
  • Lid features selectable venting or non-venting positions where the lid turns through 90° to allow for venting or non-venting of the dish

Ordering Information:

Not currently available in North America

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPetri dish, 100mm
No. per Case120
No. per Pack4
ShapeSquare
TypePetri Dish
VentSelectable
Bottom ShapeFlat
Height (Metric)20.8 mm
SterilityNon-sterile
Well Count25
Unit SizeEach

Frequently asked questions (FAQs)

What are the concentrations of the components in your BrdU Labeling reagent (Cat# 00-0103)?

Product 00-0103 contains BrdU (5-bromo-2'-deoxyuridine) and FrdU (5-fluoro-2'-deoxyuridine) at a molar ratio of 10:1. The concentrations of BrdU and FrdU in product are approximately 3 mg/mL and 0.3 mg/mL, respectively.

How long does the BrdU labeling reagent (Cat. No. 00-0103) last in cells? Can it last for 3-5 days?

The only study that we performed was in mice injected intraperitoneally with 10 uL of BrdU labeling reagent (catalog #00-0103) per gram of body weight. Incorporation of BrdU was monitored immunohistochemically only in the small intestine. Our data showed that 2-4 hours after BrdU injection in vivo was optimal for detection of staining in small intestine. Positive staining was lighter but still visible 8 hours later, but was gone after 24 hours.

With BrdU injection in mice, it is very unlikely that labeling, at least in small intestine, will last for 3-5 days. It is likely that the BrdU signal intensity as a function of time after labeling is species- and organ-specific, since the proliferation rate of the cells being labeled determines the amount of positive staining at any particular time. Also, labeling efficiency and kinetics are dependent upon the method used to label the cells in the sample with BrdU. For example, BrdU can be administered in vivo by injection, as described above, or by adding it to the animals' drinking water for several days. Labeling kinetics may vary for different types of cells in culture as well. For example, adherent cells typically label with different kinetics than cells that grow in suspension.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Rapid development of severe hyperplastic gastritis with gastric epithelial dedifferentiation in Helicobacter felis-infected IL-10(-/-) mice.
Authors:Berg DJ, Lynch NA, Lynch RG, Lauricella DM
Journal:Am J Pathol
PubMed ID:9588906
'Interleukin (IL)-10 is a potent anti-inflammatory and immune-regulatory cytokine. Mice deficient in IL-10 production (IL-10(-/-)) develop a spontaneous inflammatory bowel disease, indicating that IL-10 is an important regulator of the mucosal immune response in vivo. To study the role of IL-10 in the host response to gastric Helicobacter infection, stomachs ... More