Allyltriphenylphosphonium bromide, 99%
Allyltriphenylphosphonium bromide, 99%
Allyltriphenylphosphonium bromide, 99%
Thermo Scientific Chemicals

Allyltriphenylphosphonium bromide, 99%

CAS: 1560-54-9 | C21H20BrP | 383.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 103040250
Price (EUR)
25,80
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
25,80
Each
Chemical Identifiers
CAS1560-54-9
IUPAC Nametriphenyl(prop-2-en-1-yl)phosphanium bromide
Molecular FormulaC21H20BrP
InChI KeyFWYKRJUVEOBFGH-UHFFFAOYSA-M
SMILES[Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Argentometric>=98.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only