Search
View additional product information for Aluminon - FAQs (103060250, 103061000)
1 product FAQs found
Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.