Aminoacetaldehyde Diethyl Acetal, 99%
Aminoacetaldehyde Diethyl Acetal, 99%
Aminoacetaldehyde Diethyl Acetal, 99%
Thermo Scientific Chemicals

Aminoacetaldehyde Diethyl Acetal, 99%

CAS: 645-36-3 | C6H15NO2 | 133.19 g/mol
数量:
50 mL
250 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 103072500
価格(JPY)
-
数量:
250 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS645-36-3
IUPAC Name2,2-diethoxyethan-1-amine
Molecular FormulaC6H15NO2
InChI KeyHJKLEAOXCZIMPI-UHFFFAOYSA-N
SMILESCCOC(CN)OCC
さらに表示
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only