4'-Aminoacetophenone, 99%
4'-Aminoacetophenone, 99%
4'-Aminoacetophenone, 99%
Thermo Scientific Chemicals

4'-Aminoacetophenone, 99%

CAS: 99-92-3 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 103091000
Price (USD)
76.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.00
Save 13.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.00
Save 13.35
Each
Specifications
Assay Percent Range99%
Beilstein14, 46
CAS Max %100.0
CAS Min %98.5
CAS99-92-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only