4-Aminoantipyrine, 98%
4-Aminoantipyrine, 98%
4-Aminoantipyrine, 98%
Thermo Scientific Chemicals

4-Aminoantipyrine, 98%

4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow | CAS: 83-07-8 | C11H13N3O | 203.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 103150250
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.50
Save 8.85 (15%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.50
Save 8.85 (15%)
Each
Chemical Identifiers
CAS83-07-8
IUPAC Name4-amino-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Molecular FormulaC11H13N3O
InChI KeyRLFWWDJHLFCNIJ-UHFFFAOYSA-N
SMILESCN1N(C(=O)C(N)=C1C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with HClO4>=97.5 % (On dry substance)
Loss on drying=<1.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only