3-Aminobenzotrifluoride, 98%
3-Aminobenzotrifluoride, 98%
3-Aminobenzotrifluoride, 98%
Thermo Scientific Chemicals

3-Aminobenzotrifluoride, 98%

CAS: 98-16-8 | C7H6F3N | 161.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 103191000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-16-8
IUPAC Name3-(trifluoromethyl)aniline
Molecular FormulaC7H6F3N
InChI KeyVIUDTWATMPPKEL-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4790 to 1.4810 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to orange
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only