(4-Aminobenzoyl)hydrazide, 95%
(4-Aminobenzoyl)hydrazide, 95%
(4-Aminobenzoyl)hydrazide, 95%
Thermo Scientific Chemicals

(4-Aminobenzoyl)hydrazide, 95%

CAS: 5351-17-7 | C7H9N3O | 151.17 g/mol
Catalog NumberQuantity
1032000505 g
Catalog number 103200050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS5351-17-7
IUPAC Name4-aminobenzohydrazide
Molecular FormulaC7H9N3O
InChI KeyWPBZMCGPFHZRHJ-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point224°C to 228°C
HPLC>=94 %
Infrared spectrumConforms
Appearance (Color)White to beige
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only