RNase AWAY™ Reagent is a ready-to-use solution for eliminating RNase contamination from labware. Apply it evenly over the surface ofRead more
Have Questions?
Catalog Number
Quantity
10328011
250 mL
Catalog number 10328011
Price (MXN)
2,514.19
Each
Quantity:
250 mL
Price (MXN)
2,514.19
Each
RNase AWAY™ Reagent is a ready-to-use solution for eliminating RNase contamination from labware. Apply it evenly over the surface of glassware or plasticware to be treated and then rinse it away with distilled water. Unwanted RNase contamination are eliminated. RNase AWAY™ Reagent is nonabrasive, noncarcinogenic, and nonbiologically corrosive.
Performance and quality testing RNase AWAY™ Reagent is functionally tested for the elimination of RNase activity. No detectable RNase activity is observed.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)RNA Extraction
FormatLiquid
Quantity250 mL
Reagent TypeRNase Control Reagent
Shipping ConditionRoom Temperature
Product LineRNase Away
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
What is the recommended contact duration time of the RNase AWAY Decontamination Reagent (Cat. No. 10328011) with the according surface which has to be decontaminated, before rinsing?
There is no recommended time period for the RNase AWAY Decontamination Reagent (Cat. No. 10328011) to be in contact with the surface which has to be decontaminated. You do not need to leave the product act for a given time. Just apply it and rub with a wipe. After that you can rinse it with water.
Can I use RNAse AWAY Decontamination Reagent to get rid of DNA contamination?
RNase AWAY Decontamination Reagent is a ready-to-use solution for eliminating RNase and DNA contamination from labware. Apply it evenly over the surface of glassware or plasticware to be treated and then rinse it away with distilled water.
What quality testing is performed on your RNase AWAY Decontamination Reagent (Cat. No. 10328011)?
RNase AWAY Decontamination Reagent is functionally tested for the elimination of RNase activity and DNA. No detectable RNase activity or DNA is observed.