Interested in a bulk or custom version of this product? Request pricing.

IFNA4 Recombinant Human Protein, His Tag

Catalog number: 10336H08B25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interferon Alpha 4 (IFNA4), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications

This recombinant protein was expressed from a DNA sequence encoding the human IFNα 4a (NP_066546.1)) (Met 1-Asp 189) fused to a polyhistidine tag at the C-terminus.

Activity: The activity of this protein has not been tested.

Formulation: 20 mM Tris, 500 mM NaCl, 10% Glycerol, pH 7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: INFA4, MGC142200
Gene ID: 3441
Gene Symbol: IFNA4
Molecular Weight: 20.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Cytokines & Receptors
Protein Form: Full Length, Recombinant
Protein Name: interferon alpha 4
Protein Subtype: Interferons
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols

Recommended products