4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%
4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%
4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%
Thermo Scientific Chemicals

4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%

CAS: 1005-38-5 | C5H6ClN3S | 175.64 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 103390050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1005-38-5
IUPAC Name6-chloro-2-(methylsulfanyl)pyrimidin-4-amine
Molecular FormulaC5H6ClN3S
InChI KeyISUXMAHVLFRZQU-UHFFFAOYSA-N
SMILESCSC1=NC(N)=CC(Cl)=N1
さらに表示
Appearance (Color)Off-white to yellow to beige
Infrared spectrumConforms
GC>=96.0 %
Melting point130°C to 134°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only