3-Amino-2-chloropyridine, 96%
3-Amino-2-chloropyridine, 96%
3-Amino-2-chloropyridine, 96%
Thermo Scientific Chemicals

3-Amino-2-chloropyridine, 96%

CAS: 6298-19-7 | C5H5ClN2 | 128.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 103420100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6298-19-7
IUPAC Name2-chloropyridin-3-amine
Molecular FormulaC5H5ClN2
InChI KeyMEQBJJUWDCYIAB-UHFFFAOYSA-N
SMILESNC1=CC=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point76°C to 81°C
Infrared spectrumConforms
HPLC>=95 %
Appearance (Color)Off-white to yellow to pink
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only