IFNA5 Recombinant Human Protein (hIgG1-Fc Tag)

Catalog number: 10342H01H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

IFNA5 interferon, alpha 5 (INFA5), recombinant human protein is supplied as a lyophilized powder. It is suitable for performing cell based assays. In general, recombinant proteins are suitable for use in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding human IFNα G ( NP_002160.1 ) ( Leu 22 - Glu 189 ) fused to Fc region of human IgG1 at the N-terminus.

Activity:Measured in antiviral assays using WISH human amnion cells infected with vesicular stomatitis virus ( VSV). The EC50 for this effect is 0.16-0.8 ng/ml.

Formulation: PBS, pH 7.4.<

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

Specifications

Research Category: Drug Development, Immunology, Microbiology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: RP11-380P16.5, INFA5
Gene Symbol: IFNA5
Molecular Weight: 46.3 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Cytokines & Receptors
Protein Form: Full Length, Recombinant
Protein Name: Interferon, alpha-5
Protein Subtype: Interferons
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products