Interested in a bulk or custom version of this product? Request pricing.

SLPI Recombinant Human Protein, N-His Tag

Catalog number: 10343H07B50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Secretary Leukocyte Peptidase Inhibitor (SLPI), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the secreted form of human SLPI (P03973) (Ser 26-Ala 132) fused to a polyhistidine tag at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: 20 mM Tris, 500 mM NaCl, 10% Glycerol, pH 7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Proteases Peptidases & Inhibitors
Research Category: Immunology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: ALK1, ALP, BLPI, HUSI, HUSI-I, MPI, WAP4, WFDC4
Gene ID: 6590
Gene Symbol: SLPI
Molecular Weight: 14 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Form: Full Length, Recombinant
Protein Name: Secretory leukocyte peptidase inhibitor
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: > 84% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols