Interested in a bulk or custom version of this product? Request pricing.

IFNW1 Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10353H01H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interferon, omega-1 (IFNW1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the human IFNW1 (NP_002168.1) (Cys 24-Ser 195) fused to the Fc region of human IgG1 at the N-terminus.

Activity: Measured in an antiviral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV). The EC50 for this effect is 0.1-0.5 ng/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IFNW1, IFN-omega 1, interferon omega-1
Gene Symbol: IFNW1
Molecular Weight: 46.6 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Cytokines & Receptors
Protein Form: Full Length, Recombinant
Protein Name: interferon, omega 1
Protein Subtype: Interferons
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products