Interested in a bulk or custom version of this product? Request pricing.

TGFBR2, Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag

Catalog number: 10358H03H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Transforming Growth Factor, Beta Receptor II (TGFBR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 159) of human TGFβ receptor 2 (NP_003233.4) fused to the fused C-terminal His-tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its ability to inhibit TGF-beta1 activity on Mv-1-lu mink lung epithelial cells. The ED50 for this effect is typically 1-3 µg/ml in the presence of 1 ng/ml of recombinant human TGF-beta1.

Formulation:140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Growth Factors & Receptors
Research Category: Cardiovascular Research, Immunology, Neurobiology, Oncology, Stem Cell Research
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Gene Aliases: AAT3, FAA3, MFS2, RIIC, LDS1B, LDS2B, TAAD2, TGFR-2, TGFbeta-RII, TGFBR2
Gene ID: 7048
Gene Symbol: TGFBR2
Molecular Weight: 43.4 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Recombinant, Truncated
Protein Name: Transforming growth factor, beta receptor II
Protein Tag or Fusion: hIgG1-Fc His
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of protein in suspension buffer. Store at -5 to -30°C.

Documents

Manuals & protocols

Recommended products