Bac-to-Bac™ Vector Kit
Gibco™

Bac-to-Bac™ Vector Kit

The Bac-to-Bac™ Vector Kit contains a pFastBac™ 1 vector, as well an expression control vector, intended for use as partRead more
Have Questions?
Catalog NumberQuantity
103600141 kit
Catalog number 10360014
Price (MXN)
-
Quantity:
1 kit
The Bac-to-Bac™ Vector Kit contains a pFastBac™ 1 vector, as well an expression control vector, intended for use as part of the Bac-to-Bac™ Baculovirus Expression System (Cat. No. 10359-016), which enables the efficent production of recombinant baculovirus for expression testing in insect cells. The Bac-to-Bac™ System relies on generation of recombinant baculovirus by site-specific transposition in E. colirather than homologous recombination in insect cells (Figure 1). This system features:

• Time-saving expression bacmid. With Bac-to-Bac™, the expression cassette of the pFastBac™ vector recombines with the parent bacmid in DH10Bac™ E. coli Competent Cells (not included with this vector kit) to form an expression bacmid. The bacmid is then transfected into insect cells for production of recombinant baculovirus particles.
• Easy colony screening. The parent bacmid in DH10Bac™ E. colicontains a segment of the lacZα gene. The lacZα gene is disrupted upon transposition of the expression cassette into the bacmid allowing for blue/white selection of recombinants. This makes identification of recombinant colonies easy.
The Bac-to-Bac™ Baculovirus Expression System is designed for fast, small-scale production of recombinant baculovirus. The pFastBac™ 1 vector provided by this kit offers the strong polyhedrin promoter for protein expression and a large multiple cloning site for simplified cloning.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeVector Kit
Quantity1 kit
VectorpFastBac
Cloning MethodRestriction Enzyme/MCS
Product LineBac-to-Bac
PromoterPolyhedrin
Protein TagUntagged
Unit SizeEach
Contents & Storage
Contains 10 μg of pFastBac™1 vector and pFastBac™ 1-Gus control vector.

Store at -20°C.

Frequently asked questions (FAQs)

I cannot grow this white colony in liquid culture. What should I do?

The concentration of gentamicin might be too high. Try lowering the amount to 5 µg/mL and try adding more of the colony to the culture medium.

What has happened when I see blue colonies? How about colonies which are blue in the center and white on the edges?

In the case of a blue colony, the E. coli has the bacmid and the plasmid in it, allowing the cells to survive the selection process. However, because the transposition has not occurred, the LacZ gene is not disrupted. For bulls-eye colonies, this indicates that the transposition took place when the colony was growing. Re-streaking for an isolated clone from the white portion of the mixed colony should yield some colonies where transposition occurred.

I'm getting mostly white/wild-type plaques instead of blue/recombinant plaques. What am I doing wrong?

This is typically an indication of poor homologous recombination. Check the plasmid/linear DNA ratio you used. If there are some blue plaques, however, expand those viruses and check for their protein. In our experience, they are correct, even if they were in relatively low abundance.

I've infected my cells and see large polyhedra in one cell and smaller polyhedra (more numerous) in a neighboring cell. Is this normal?

Yes, cells are infected with wild-type virus individually and will develop polyhedra at different rates until all the cells in the flask are infected. The polyhedra in cells will form in approximately 3-4 days, differing in size and number until they reach their maximum capacity and burst the cell, releasing tiny particles of virus into the medium.

I'm worried that I am not getting plaques. How many days does it take to see plaques and what size are they typically?

Normally, very small white dots show up about 5-7 days and 1 mm plaques show up around day 10. Plaques can vary in size from 1 mm to 4 mm.

Citations & References (12)

Citations & References
Abstract
Cytoskeletal changes regulated by the PAK4 serine/threonine kinase are mediated by LIM kinase 1 and cofilin.
Authors: Dan C; Kelly A; Bernard O; Minden A;
Journal:J Biol Chem
PubMed ID:11413130
'PAK4 is the most recently identified member of the PAK family of serine/threonine kinases. PAK4 differs from other members of the PAK family in sequence and in many of its functions. Previously, we have shown that an important function of this kinase is to mediate the induction of filopodia in ... More
Allocation of helper T-cell epitope immunodominance according to three-dimensional structure in the human immunodeficiency virus type I envelope glycoprotein gp120.
Authors: Dai G; Steede N K; Landry S J;
Journal:J Biol Chem
PubMed ID:11551929
'The specificity and intensity of CD4(+) helper T-cell responses determine the effectiveness of immune effector functions. Promiscuously immunodominant helper T-cell epitopes in the human immunodeficiency virus (HIV) envelope glycoprotein gp120 could be important in the development of broadly protective immunity, but the underlying mechanisms of immunodominance and promiscuity remain poorly ... More
Interaction of Fibroblast Growth Factor Receptor 3 and the Adapter Protein SH2-B. A ROLE IN STAT5 ACTIVATION.
Authors: Kong Monica; Wang Ching S; Donoghue Daniel J;
Journal:J Biol Chem
PubMed ID:11827956
'Fibroblast growth factor receptor 3 (FGFR3) influences a diverse array of biological processes, including cell growth, differentiation, and migration. Activating mutations in FGFR3 are associated with multiple myeloma, cervical carcinoma, and bladder cancer. To identify proteins that interact with FGFR3 and which may mediate FGFR3-dependent signaling, a yeast two-hybrid screen ... More
Functional differences between the human ATP-dependent nucleosome remodeling proteins BRG1 and SNF2H.
Authors: Aalfs J D; Narlikar G J; Kingston R E;
Journal:J Biol Chem
PubMed ID:11435432
'ATP-dependent nucleosome remodeling complexes can be grouped into several classes that may differ in their biochemical remodeling activities and biological roles. Although there are a number of biochemical studies of each class of remodeler, there are very little data directly comparing the biochemical activities of remodelers from different classes. We ... More
PIAS1 and PIASxalpha function as SUMO-E3 ligases toward androgen receptor and repress androgen receptor-dependent transcription.
Authors:Nishida T, Yasuda H,
Journal:J Biol Chem
PubMed ID:12177000
The androgen receptor (AR) has been shown to be modified by SUMO-1, a ubiquitin-like protein. Recently we showed that PIAS family proteins function as SUMO-E3 ligases. Here we provide evidence that PIAS1 and PIASxalpha act as specific SUMO-E3 ligases for the AR. PIAS1 and PIASxalpha but not PIAS3 or PIASxbeta ... More