Donor Bovine Serum with Iron, New Zealand Origin
Gibco™

Donor Bovine Serum with Iron, New Zealand Origin

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Have Questions?
카탈로그 번호수량
10371029500 mL
카탈로그 번호 10371029
제품 가격(KRW)
134,000
Each
수량:
500 mL
제품 가격(KRW)
134,000
Each

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Origin: New Zealand.
  • Endotoxin level: ≤10EU/mL
  • Collected from cattle 12-36 months of age

Cell Culture, Mammalian Cell Culture

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
사양
엔도톡신 농도≤ 10 EU/mL
제품라인Gibco
배송 조건Frozen
종Cattle/Bovine
Age12 to 36 months
원산지New Zealand
형태Liquid
형식Bottle
제품 유형Fetal Bovine Serum
수량500 mL
Serum TreatmentStandard (Sterile-filtered)
멸균Sterile
Unit SizeEach
구성 및 보관
Storage conditions: ≤-10°C
Shipping conditions: Frozen

자주 묻는 질문(FAQ)

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.