Interested in a bulk or custom version of this product? Request pricing.

FCGR2A (CD32a) Recombinant Human Protein, His Tag

Catalog number: 10374H08H50

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human FCGR2A precursor (AAA35827.1) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized recombinant human FCGR2A at 10 µg/ml(100 µl/well) can bind human IgG2 with a linear range of 12.5-200 ng/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Immunology, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, MGC23887, MGC30032
Gene ID: 2212
Gene Symbol: FCGR2A
Molecular Weight: 22 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: Fc fragment of IgG, low affinity IIa, receptor (CD32)
Protein Subtype: Fc Receptors, Glycoproteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols