Micro Length Non-Filtered Pipette Tips, 0.1-10 μL
Thermo Scientific™

Micro Length Non-Filtered Pipette Tips, 0.1-10 μL

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatSterility
104-QBaggedNon-sterile
T104R-QHinged RackNon-sterile
T104RL-QRefillNon-sterile
T104RLS-QRefillSterile
T104RS-QHinged RackSterile
Catalog number 104-Q
Price (MXN)
-
Format:
Bagged
Sterility:
Non-sterile
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
DescriptionQSP 10, Non-Filtered
Length (Metric)3.1 cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatBagged
Product LineQSP
SterilityNon-sterile
Tip StyleMicro Point
Volume (Metric)10 μL
Unit SizeCase of 10000