Interested in a bulk or custom version of this product? Request pricing.

CD4 Recombinant Human Protein, hIgG1-Fc.His Tag

Catalog number: 10400H03H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Manufactured by Sino Biological (China); Distributed by Life Technologies.

Cluster of Differentiation 4 (CD4) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in the study of protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 390) of human CD4 (NP_000607.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus.

N-terminal Sequence Analysis: Lys26.

Activity: measured by its binding ability in a functional ELISA. Immobilized human CD4-His at 2 µg/well has been shown to bind biotinylated HIV-1 GP120 with a linear range of 0.078-1.25 µg/mL.

Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80.

Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: expires one year from date of receipt when stored as instructed.

Background
Human T-cell surface glycoprotein CD4, also known as T-cell surface antigen T4/Leu-3, is a single-pass type I membrane protein. CD4 contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like Vtype (immunoglobulin-like) domain. CD4 is a glycoprotein expressed on the surface of T helper cells, regulatory T cells, monocytes, macrophages, and dendritic cells. CD4 amplifies the signal generated by the TCR. CD4 interacts directly with MHC class II molecules on the surface of the antigen presenting cell via its extracellular domain. CD4 is a primary receptor used by HIV-1 to gain entry into host T cells. HIV infection leads to a progressive reduction of the number of T cells possessing CD4 receptors. Therefore, medical professionals refer to the CD4 count to decide when to begin treatment for HIV-infected patients.

You may also be interested in the following CD4 products:
Primary Antibodies
Gene Expression Assays
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Drug Development, Immunology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CD4mut
Gene ID: 920
Gene Symbol: CD4
Molecular Weight: 68.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: CD4 molecule
Protein Subtype: Glycoproteins
Protein Tag or Fusion: His Tag, IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols