2-Amino-2-methyl-1,3-propanediol, 99%
2-Amino-2-methyl-1,3-propanediol, 99%
2-Amino-2-methyl-1,3-propanediol, 99%
Thermo Scientific Chemicals

2-Amino-2-methyl-1,3-propanediol, 99%

CAS: 115-69-5 | C4H11NO2 | 105.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 104051000
Price (USD)
76.65
Online exclusive
85.10
Save 8.45
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
76.65
Online exclusive
85.10
Save 8.45
Each
Chemical Identifiers
CAS115-69-5
IUPAC Name2-amino-2-methylpropane-1,3-diol
Molecular FormulaC4H11NO2
InChI KeyUXFQFBNBSPQBJW-UHFFFAOYSA-N
SMILESCC(N)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point>=100°C
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only