2-(Aminomethyl)pyridine, 99%
2-(Aminomethyl)pyridine, 99%
2-(Aminomethyl)pyridine, 99%
Thermo Scientific Chemicals

2-(Aminomethyl)pyridine, 99%

CAS: 3731-51-9 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
Packaging:
Glass bottle
Catalog number 104075000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3731-51-9
IUPAC Name1-(pyridin-2-yl)methanamine
Molecular FormulaC6H8N2
InChI KeyWOXFMYVTSLAQMO-UHFFFAOYSA-N
SMILESNCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5430 to 1.5470 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only