Interested in a bulk or custom version of this product? Request pricing.

TNFSF10 Recombinant Human Protein

Catalog number: 10409HNAE25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tumor Necrosis (Ligand) Receptor Superfamily Member 10 (TNFSF10), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications

This recombinant protein was expressed from a DNA sequence encoding the human TNFSF10 (NP_003801.1) (Val 114-Gly 281) fused to an initial Met.

Activity:
1. Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D. The ED50 for this effect is typically 4-8 ng/ml.

2. Measured in a cytotoxicity assay using NCI-H460 cells in the presence of the metabolic inhibitor actinomycin D. The ED50 for this effect is typically 0.2-0.8 ng/ml.

Formulation: Lyophilized in 40 mM Tris 0.3M NaCl PH7.05% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Apoptosis, Oncology, Signal Transduction
Expression: E. coli
Form: Lyophilized
Gene Aliases: APO2L, Apo-2L, CD253, TL2, TRAIL
Gene Symbol: TNFSF10
Molecular Weight: 19.6 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: CD Proteins, Cytokines & Receptors
Protein Name: tumor necrosis (ligand) receptor superfamily, member 10
Protein Subtype: TNF (Tumor Necrosis Factor)
Protein Tag or Fusion: Untagged
Purity: ≥ 95% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products