N-Aminomorpholine, 95%
N-Aminomorpholine, 95%
N-Aminomorpholine, 95%
Thermo Scientific Chemicals

N-Aminomorpholine, 95%

CAS: 4319-49-7 | C4H10N2O | 102.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1041100505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 104110050
Price (EUR)
44,90
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
44,90
Each
Chemical Identifiers
CAS4319-49-7
IUPAC Namemorpholin-4-amine
Molecular FormulaC4H10N2O
InChI KeyMKQLBNJQQZRQJU-UHFFFAOYSA-N
SMILESNN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4770 to 1.4800 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only