2-Aminonicotinic acid, 98%
2-Aminonicotinic acid, 98%
2-Aminonicotinic acid, 98%
Thermo Scientific Chemicals

2-Aminonicotinic acid, 98%

CAS: 5345-47-1 | C6H6N2O2 | 138.13 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 104160250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5345-47-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only