2-Amino-5-nitropyridine, 99%
2-Amino-5-nitropyridine, 99%
2-Amino-5-nitropyridine, 99%
Thermo Scientific Chemicals

2-Amino-5-nitropyridine, 99%

CAS: 4214-76-0 | C5H5N3O2 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
250 g
Packaging:
View all
Glass Bottle
Catalog number 104232500
Price (USD)
435.00
Each
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
435.00
Each
Chemical Identifiers
CAS4214-76-0
IUPAC Name5-nitropyridin-2-amine
Molecular FormulaC5H5N3O2
InChI KeyUGSBCCAHDVCHGI-UHFFFAOYSA-N
SMILESNC1=CC=C(C=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point186°C to 192°C
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only