4-Aminophenol, 97%
4-Aminophenol, 97%
4-Aminophenol, 97%
Thermo Scientific Chemicals

4-Aminophenol, 97%

CAS: 123-30-8 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 104272500
Price (USD)
45.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
45.00
Each
Chemical Identifiers
CAS123-30-8
IUPAC Name4-aminophenol
Molecular FormulaC6H7NO
InChI KeyPLIKAWJENQZMHA-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to beige to light brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only