4-Aminophenyl sulfone, 97%
4-Aminophenyl sulfone, 97%
4-Aminophenyl sulfone, 97%
Thermo Scientific Chemicals

4-Aminophenyl sulfone, 97%

CAS: 80-08-0 | C12H12N2O2S | 248.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 104361000
Price (EUR)
47,50
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
47,50
Each
Chemical Identifiers
CAS80-08-0
IUPAC Name4-(4-aminobenzenesulfonyl)aniline
Molecular FormulaC12H12N2O2S
InChI KeyMQJKPEGWNLWLTK-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Melting point173°C to 178°C
Titration with HClO4>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only