4-Aminosalicylic acid, 99%
4-Aminosalicylic acid, 99%
4-Aminosalicylic acid, 99%
Thermo Scientific Chemicals

4-Aminosalicylic acid, 99%

CAS: 65-49-6 | C7H7NO3 | 153.14 g/mol
Quantity:
5 g
100 g
500 g
Catalog number 104620050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS65-49-6
IUPAC Name4-amino-2-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyWUBBRNOQWQTFEX-UHFFFAOYSA-N
SMILESNC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=98.5 %
Appearance (Color)Beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only