Interested in a bulk or custom version of this product? Request pricing.

MMP3 Recombinant Human Protein

Catalog number: 10467HNAE25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Matrix Metallopeptidase 3 (MMP3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human MMP3 (AAA36321.1) N-terminal fragment (Tyr 18-Thr 272).

Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2, AnaSpec, Catalog # 27114.
The specific activity is >300 pmoles/min/µg.

Formulation: Lyophilized in 50 mM Tris 10 mMCaCL2 1uM ZnCL2 50 mM Nacl 0.5%Brij35 PH=7.05% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Angiogenesis, Oncology, Other Research
Expression: E. coli
Form: Lyophilized
Gene Aliases: CHDS6, MMP-3, SL-1, STMY, STMY1, STR1
Gene Symbol: MMP3
Molecular Weight: 29 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Name: matrix metallopeptidase 3
Protein Subtype: Proteases
Protein Tag or Fusion: Untagged
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols