TrackIt™ Cyan/Orange Loading Buffer
Invitrogen™

TrackIt™ Cyan/Orange Loading Buffer

TrackIt loading buffers are 6X solutions used for easy loading and tracking of DNA samples in agarose gels. They contain자세히 알아보기
Have Questions?
카탈로그 번호수량
104820283 x 0.5 mL
카탈로그 번호 10482028
제품 가격(KRW)
71,000
온라인 행사
Ends: 30-Sep-2026
81,000
할인액 10,000 (12%)
Each
수량:
3 x 0.5 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
71,000
온라인 행사
Ends: 30-Sep-2026
81,000
할인액 10,000 (12%)
Each
TrackIt loading buffers are 6X solutions used for easy loading and tracking of DNA samples in agarose gels. They contain two tracking dyes: one that runs behind the sample and one that runs ahead of the sample. The tracking dyes serve as visual markers of migration during electrophoresis and indicate when maximum resolution of the DNA fragments has been achieved. Additionally, visualization of DNA bands is not obscured by the tracking dyes because they run outside the limits of most DNA samples.TrackIt Cyan/Orange Loading Buffer is recommended for DNA fragments between 50 bp and 10 Kb.TrackIt Cyan/Yellow Loading Buffer is recommended for DNA fragments between 10 bp and 1 Kb.
For Research Use Only. Not for use in diagnostic procedures.
사양
버퍼Sample Loading Buffers
농도6X
젤 호환성Agarose Gels
수량3 x 0.5 mL
샘플 로딩 부피0.5 mL
유통 기한6 Months
배송 조건Approved for shipment at Room Temperature or on Wet or Dry Ice
젤 유형Agarose
라벨 또는 염료Orange G, Xylene Cyanol, Xylene Cyanol FF
제품라인TrackIt
제품 유형Loading Buffer
Unit SizeEach
구성 및 보관
• 3 x 0.5 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.